Benzyl acetate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W213500-10KG-K W213500-1KG-K W213500-25KG-K
SIALW213500-10KG-KEA 249 EUR
SIALW213500-10KG-K SIALW213500-1KG-K SIALW213500-25KG-K
Benzyl acetate, Sigma-Aldrich®
Benzyl acetate
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 206 °C (1013 hPa)
Melting Pt: –51 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00008712
CAS Number: 140-11-4
EINECS: 205-399-7
Merck Index: 13,01125