Benzyl acetate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W213519-100G-K W213519-1KG-K W213519-5KG-K
SIALW213519-100G-KEA 151 EUR
SIALW213519-100G-K SIALW213519-1KG-K SIALW213519-5KG-K
Benzyl acetate, Sigma-Aldrich®
Benzyl acetate
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 206
Melting Pt: –51 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 95
Storage Temperature: Ambient
MDL Number: MFCD00008712
CAS Number: 140-11-4
EINECS: 205-399-7
Merck Index: 13,01125