Ethyl phenylacetate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W245208-10KG-K
W245208-1KG-K
W245208-5KG-K
SIALW245208-10KG-KEA
592
EUR
SIALW245208-10KG-K
SIALW245208-1KG-K
SIALW245208-5KG-K
Ethyl phenylacetate, Sigma-Aldrich®
Ethyl phenylacetate
Formula:
C₁₀H₁₂O₂ MW: 164.2 g/mol Boiling Pt: 226 °C (1013 hPa) Melting Pt: –29 °C Density: 1.03 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009178 CAS Number: 101-97-3 EINECS: 202-993-8 Merck Index: 13,03872 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...