3-Phenylpropionic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3-Phenylpropanoic acid, Hydrocinnamic acid
M-Clarity™ quality level = MQ100
135232-100G 135232-5G
SIAL135232-100GEA 53.3 EUR
SIAL135232-100G SIAL135232-5G
3-Phenylpropionic acid, Sigma-Aldrich®
3-Phenylpropionic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 46…47 °C
Density: 1.071 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002771
CAS Number: 501-52-0
EINECS: 207-924-5
Merck Index: 13,04805