Diphenylacetic acid ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
A11489.22
A11489.36
A11489.22EA
60.7
EUR
A11489.22
A11489.36
Diphenylacetic acid ≥99%
Diphenylacetic acid
Purity: 99%
CAS No.: 117-34-0
Molecular Formula: C14H12O2
Formula Weight: 212.25
Odour: Odourless
pH: 3.8
Melting Point: 146-148°
Boiling Point: 195°/25mm
Beilstein Reference: 1910978
Merck Reference: 14,3316
MDL No.: MFCD00004251
Formula:
C₁₄H₁₂O₂ Boiling Pt: 195 °C (25 mmHg) Melting Pt: 146…148 °C Density: 1.258 g/cm³ (15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004251 CAS Number: 117-34-0 EINECS: 204-185-0 Merck Index: 14,3316 |