(3,4-Dimethoxyphenyl)acetic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: Homoveratric acid, 3,4-Dimethoxyphenylacetic acid
M-Clarity™ quality level = MQ100
D135909-100G D135909-25G
SIALD135909-100GEA 69.8 EUR
SIALD135909-100G SIALD135909-25G
(3,4-Dimethoxyphenyl)acetic acid, Sigma-Aldrich®
(3,4-Dimethoxyphenyl)acetic acid
Formula: (C₆H₃(OCH₃)₂)CH₂COOH
MW: 196.2 g/mol
Boiling Pt: 327 °C (1013 hPa)
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00004335
CAS Number: 93-40-3
EINECS: 202-244-5