(2-Hydroxyphenyl)acetic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Hydroxyphenylacetic acid
M-Clarity™ quality level = MQ100
H49804-10G H49804-25G
SIALH49804-10GEA 56.7 EUR
SIALH49804-10G SIALH49804-25G
(2-Hydroxyphenyl)acetic acid, Sigma-Aldrich®
(2-Hydroxyphenyl)acetic acid
Formula: (C₆H₄OH)CH₂COOH
MW: 152.15 g/mol
Boiling Pt: 328
Melting Pt: 145…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00004323
CAS Number: 614-75-5
EINECS: 210-393-2