(3-Hydroxyphenyl)acetic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3-Hydroxyphenylacetic acid
M-Clarity™ quality level = MQ100
H49901-25G H49901-5G
SIALH49901-25GEA 125 EUR
SIALH49901-25G SIALH49901-5G
(3-Hydroxyphenyl)acetic acid, Sigma-Aldrich®
(3-Hydroxyphenyl)acetic acid
Formula: (C₆H₄(OH))CH₂COOH
MW: 152.15 g/mol
Boiling Pt: 349 °C (1013 hPa)
Melting Pt: 129…133 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004337
CAS Number: 621-37-4
EINECS: 210-684-4