(2,5-Difluorophenyl)acetic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2,5-Difluorophenylacetic acid
M-Clarity™ quality level = MQ100
290424-1G 290424-5G
SIAL290424-1GEA 33.6 EUR
SIAL290424-1G SIAL290424-5G
(2,5-Difluorophenyl)acetic acid, Sigma-Aldrich®
(2,5-Difluorophenyl)acetic acid
Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 122…128 °C
MDL Number: MFCD00010000
CAS Number: 85068-27-5
EINECS: 285-288-8