(3,4-Difluorophenyl)acetic acid, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 3,4-Difluorophenylacetic acid

M-Clarity™ quality level = MQ100

290432-1G 290432-5G
SIAL290432-1GEA 26.1 EUR
SIAL290432-1G SIAL290432-5G
(3,4-Difluorophenyl)acetic acid, Sigma-Aldrich®
(3,4-Difluorophenyl)acetic acid
Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 40…42 °C
MDL Number: MFCD00010002
CAS Number: 658-93-5
EINECS: 211-527-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR