6-Bromo-1-hexene, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

247219-1G 247219-5G
SIAL247219-1GEA 22 EUR
SIAL247219-1G SIAL247219-5G
6-Bromo-1-hexene, Sigma-Aldrich®
6-Bromo-1-hexene
Formula: C₆H₁₁Br
MW: 163.06 g/mol
Boiling Pt: 34…36 °C (0.05 mmHg)
Density: 1.2169 g/cm³ (20 °C)
MDL Number: MFCD00000269
CAS Number: 2695-47-8
EINECS: 220-267-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR