8-Bromo-1-octene, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

252301-1G 252301-5G
SIAL252301-1GEA 28.5 EUR
SIAL252301-1G SIAL252301-5G
8-Bromo-1-octene, Sigma-Aldrich®
8-Bromo-1-octene
Formula: C₈H₁₅Br
MW: 191.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000275
CAS Number: 2695-48-9
EINECS: 220-268-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR