cis-4-Heptenal, Sigma-Aldrich®
Supplier: Merck
Synonyms: cis-4-Heptenal, Cis-4-heptenal, (Z)-4-Heptenal
M-Clarity™ quality level = MQ300
W328901-100G-K W328901-1KG-K
SIALW328901-100G-KEA 432 EUR
SIALW328901-100G-K SIALW328901-1KG-K
cis-4-Heptenal, Sigma-Aldrich®
cis-4-Heptenal
Formula: C₇H₁₂O
MW: 112.17 g/mol
Boiling Pt: 46 °C (1013 hPa)
Density: 0.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014682
CAS Number: 6728-31-0
EINECS: 229-779-7
UN: 1989
ADR: 3,III