trans-2-Hexenal, Sigma-Aldrich®
Supplier: Merck
Synonyms: (E)-2-Hexenal, trans-2-Hexenal
M-Clarity™ quality level = MQ100
132659-25G 132659-5G
SIAL132659-25GEA 44.1 EUR
SIAL132659-25G SIAL132659-5G
trans-2-Hexenal, Sigma-Aldrich®
trans-2-Hexenal
Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 147 °C (1013 hPa)
Density: 0.846 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007008
CAS Number: 6728-26-3
EINECS: 229-778-1
UN: 1989
ADR: 3,III