trans-2-Hexenal, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(E)-2-Hexenal, trans-2-Hexenal
M-Clarity™ quality level = MQ300
W256005-1KG-K
W256005-20KG-K
W256005-4KG-K
SIALW256005-1KG-KEA
160
EUR
SIALW256005-1KG-K
SIALW256005-20KG-K
SIALW256005-4KG-K
trans-2-Hexenal, Sigma-Aldrich®
trans-2-Hexenal
Formula:
C₆H₁₀O MW: 98.14 g/mol Boiling Pt: 147 °C (1013 hPa) Density: 0.846 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007008 CAS Number: 6728-26-3 EINECS: 229-778-1 UN: 1989 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...