Ethyl vinyl ketone, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Penten-3-one

M-Clarity™ quality level = MQ200

E51309-25G E51309-5G
SIALE51309-25GEA 249 EUR
SIALE51309-25G SIALE51309-5G
Ethyl vinyl ketone, Sigma-Aldrich®
Ethyl vinyl ketone
Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 38 °C (1013 hPa)
Density: 0.8524 g/cm³ (15 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009316
CAS Number: 1629-58-9
EINECS: 216-624-3
UN: 3286
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR