Acrylamide, 2x crystallised
Supplier: BIOMOL RESEARCH LABORATORIES
BMOL50073-1EA 0 EUR
BMOL50073-1
Acrylamide, 2x crystallised
Acrylamide
Formula: C₃H₅NO
MW: 71,08 g/mol
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 84,5 °C
Density: 1,122 g/cm³ (20 °C)
Flash Pt: 138 °C
MDL Number: MFCD00008032
UN: 2074
ADR: 6.1,III
https://si.vwr.com/stibo/web/std.lang.all/13/21/2301321.jpg