Acrylamide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Prop-2-enamide
MQ200
01696-100G 01696-1KG 01696-500G
SIAL01696-100GEA 112 EUR
SIAL01696-100G SIAL01696-1KG SIAL01696-500G
Acrylamide, Sigma-Aldrich®
Acrylamide (monomer)
Formula: C₃H₅NO
MW: 71.08 g/mol
Boiling Pt: 232
Melting Pt: 84.5
Density: 1.122 g/cm³ (20 °C)
Flash Pt: 138
Storage Temperature: Refrigerator
MDL Number: MFCD00008032
CAS Number: 79-06-1
EINECS: 201-173-7
UN: 2074
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7687b7f17a8cdf31c559a33fd5d4021a2e50a40a&vid=fecebe0c34b037788df119c11bfc9637d78cda0d&prid=web&clid=SAPDAM