Ethyl oleate, Sigma-Aldrich®

Supplier: Merck

SIAL75100-1LEA 114 EUR
SIAL75100-1L
Ethyl oleate, Sigma-Aldrich®
Ethyl oleate
Formula: C₂₀H₃₈O₂
MW: 310,52 g/mol
Boiling Pt: 386 °C (1013 hPa)
Melting Pt: –32 °C
Density: 0,87 g/cm³ (20 °C)
Flash Pt: 174 °C
MDL Number: MFCD00009579
CAS Number: 111-62-6
Merck Index: 12,06965

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR