Diethyl maleate, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Maleic acid diethyl ester, Diethyl (Z)-2-butenedioate

M-Clarity™ quality level = MQ300

W505005-10KG W505005-1KG W505005-SAMPLE
SIALW505005-10KGEA 720 EUR
SIALW505005-10KG SIALW505005-1KG SIALW505005-SAMPLE
Diethyl maleate, Sigma-Aldrich®
Diethyl maleate
Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 225
Melting Pt: –11 °C
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 93
Storage Temperature: Ambient
MDL Number: MFCD00009191
CAS Number: 141-05-9
EINECS: 205-451-9
Merck Index: 13,03150

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR