Diethyl fumarate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Fumaric acid diethyl ester
M-Clarity™ quality level = MQ200
D95654-100G D95654-500G D95654-5G
SIALD95654-100GEA 124 EUR
SIALD95654-100G SIALD95654-500G SIALD95654-5G
Diethyl fumarate, Sigma-Aldrich®
Diethyl fumarate
Formula: C₈H₁₂O₄
Boiling Pt: 219 °C (1013 hPa)
Melting Pt: 1…2 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00064455
CAS Number: 623-91-6
EINECS: 210-819-7