Dimethyl fumarate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Fumaric acid dimethyl ester

MQ200

242926-25G 242926-100G
SIAL242926-25GEA 28.9 EUR
SIAL242926-25G SIAL242926-100G
Dimethyl fumarate, Sigma-Aldrich®
Dimethyl fumarate
Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 192
Melting Pt: 102
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064438
CAS Number: 624-49-7
EINECS: 210-849-0
Merck Index: 13,04308

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR