4-Pentenoic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Allylacetic acid, Pent-4-enoic acid
M-Clarity™ quality level = MQ200
245925-25G
245925-5G
SIAL245925-25GEA
150
EUR
SIAL245925-25G
SIAL245925-5G
4-Pentenoic acid, Sigma-Aldrich®
4-Pentenoic acid
Formula:
C₅H₈O₂ MW: 100.12 g/mol Boiling Pt: 187 °C (1013 hPa) Melting Pt: –18 °C Density: 0.98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004408 CAS Number: 591-80-0 EINECS: 209-732-7 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...