Oleic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: (9Z)-9-Octadecenoic acid
M-Clarity™ quality level = MQ200
364525-1L 364525-4L
SIAL364525-1LEA 61.8 EUR
SIAL364525-1L SIAL364525-4L
Oleic acid, Sigma-Aldrich®
Oleic acid
Formula: C₁₈H₃₄O₂
MW: 282.47 g/mol
Boiling Pt: 286
Melting Pt: 16.7
Density: 0.8946 g/cm³ (20 °C)
Flash Pt: 180
Storage Temperature: Freezer
MDL Number: MFCD00064242
CAS Number: 112-80-1
EINECS: 204-007-1
Merck Index: 13,06898
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aecc5dce4d0318386a45d7adb7c5822631010a&vid=892a2df4ecab3679b06983b84cd27ad334ef961f&prid=web&clid=SAPDAM