Oleic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: (9Z)-9-Octadecenoic acid

MQ200

O1008-5G O1008-1G O1008-25G
SIALO1008-5GEA 153 EUR
SIALO1008-5G SIALO1008-1G SIALO1008-25G
Oleic acid, Sigma-Aldrich®
Oleic acid
Formula: C₁₈H₃₄O₂
MW: 282.47 g/mol
Boiling Pt: 286
Melting Pt: 16.7
Density: 0.8946 g/cm³ (20 °C)
Flash Pt: 180
Storage Temperature: Freezer
MDL Number: MFCD00064242
CAS Number: 112-80-1
EINECS: 204-007-1
Merck Index: 13,06898
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aecc5dce4d0318386a45d7adb7c5822631010a&vid=892a2df4ecab3679b06983b84cd27ad334ef961f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR