Oleic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(9Z)-9-Octadecenoic acid
MQ200
O1383-1G
O1383-25G
O1383-5G
SIALO1383-1GEA
70.8
EUR
SIALO1383-1G
SIALO1383-25G
SIALO1383-5G
Oleic acid, Sigma-Aldrich®
Oleic acid
Formula:
C₁₈H₃₄O₂ MW: 282.47 g/mol Boiling Pt: 286 Melting Pt: 16.7 Density: 0.8946 g/cm³ (20 °C) Flash Pt: 180 Storage Temperature: Freezer |
MDL Number:
MFCD00064242 CAS Number: 112-80-1 EINECS: 204-007-1 Merck Index: 13,06898 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...