Oleic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: (9Z)-9-Octadecenoic acid
M-Clarity™ quality level = MQ300
W281506-10KG-K W281506-1KG-K W281506-20KG-K W281506-4KG-K
SIALW281506-10KG-KEA 310 EUR
SIALW281506-10KG-K SIALW281506-1KG-K SIALW281506-20KG-K SIALW281506-4KG-K
Oleic acid, Sigma-Aldrich®
Oleic acid
Formula: C₁₈H₃₄O₂
MW: 282.47 g/mol
Boiling Pt: 286
Melting Pt: 16.7
Density: 0.8946 g/cm³ (20 °C)
Flash Pt: 180
Storage Temperature: Freezer
MDL Number: MFCD00064242
CAS Number: 112-80-1
EINECS: 204-007-1
Merck Index: 13,06898
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aecc5dce4d0318386a45d7adb7c5822631010a&vid=892a2df4ecab3679b06983b84cd27ad334ef961f&prid=web&clid=SAPDAM