Maleic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
M0375-1KG
M0375-25KG
M0375-500G
M0375-100G
SIALM0375-1KGEA
72.4
EUR
SIALM0375-1KG
SIALM0375-25KG
SIALM0375-500G
SIALM0375-100G
Maleic acid, Sigma-Aldrich®
Maleic acid
Formula:
C₄H₄O₄ MW: 116.07 g/mol Boiling Pt: 256 °C (1013 hPa) Melting Pt: 130…131 °C Density: 1.59 g/cm³ (20 °C) Flash Pt: 127 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00063177 CAS Number: 110-16-7 EINECS: 203-742-5 UN: 3261 ADR: 8,III Merck Index: 13,05726 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...