Citraconic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Methyl-2-butenedioic acid

M-Clarity™ quality level = MQ200

C82604-100G C82604-5G
SIALC82604-100GEA 169 EUR
SIALC82604-100G SIALC82604-5G
Citraconic acid, Sigma-Aldrich®
Citraconic acid
Formula: C₅H₆O₄
MW: 130.1 g/mol
Boiling Pt: 336
Melting Pt: 98…99 °C
Density: 1.617 g/cm³ (20 °C)
MDL Number: MFCD00078086
CAS Number: 498-23-7
EINECS: 207-858-7
Merck Index: 13,02345

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR