Maleic anhydride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
2,5-Furanedione
M-Clarity™ quality level = MQ200
63200-100G-F
63200-500G-F
SIAL63200-100G-FEA
34.6
EUR
SIAL63200-100G-F
SIAL63200-500G-F
Maleic anhydride, Sigma-Aldrich®
Maleic anhydride
Formula:
C₄H₂O₃ MW: 98.06 g/mol Boiling Pt: 200 °C (1013 hPa) Melting Pt: 51…53 °C Density: 1.48 g/cm³ (20 °C) Flash Pt: 103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005518 CAS Number: 108-31-6 EINECS: 203-571-6 UN: 2215 ADR: 8,III Merck Index: 13,05727 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...