Citraconic anhydride, Sigma-Aldrich®
Supplier: Merck
Synonyms:
3-Methylfuran-2,5-dione
M-Clarity™ quality level = MQ200
125318-100G
125318-25G
125318-500G
SIAL125318-100GEA
76.7
EUR
SIAL125318-100G
SIAL125318-25G
SIAL125318-500G
Citraconic anhydride, Sigma-Aldrich®
Citraconic anhydride
Formula:
C₅H₄O₃ MW: 112.08 g/mol Boiling Pt: 213 °C (1013 hPa) Melting Pt: 7…8 °C Density: 1.243 g/cm³ (20 °C) Flash Pt: 101 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005522 CAS Number: 616-02-4 EINECS: 210-459-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...