10-Undecen-1-ol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Undecen-11-ol, 10-Undecen-1-ol
M-Clarity™ quality level = MQ200
U2008-100G U2008-25G
SIALU2008-100GEA 133 EUR
SIALU2008-100G SIALU2008-25G
10-Undecen-1-ol, Sigma-Aldrich®
10-Undecen-1-ol
Formula: C₁₁H₂₂O
MW: 170.3 g/mol
Boiling Pt: 75…80 °C (0.05 mmHg)
Melting Pt: –1 °C
Density: 0.86 g/cm³ (0 °C)
MDL Number: MFCD00004750
CAS Number: 112-43-6
EINECS: 203-971-0