4-Penten-1-ol, Sigma-Aldrich®

Supplier: Merck

Synonyms: Pent-4-en-1-ol, 1-Penten-5-ol, 4-Penten-1-ol

M-Clarity™ quality level = MQ200

111279-10G 111279-1G 111279-50G
SIAL111279-10GEA 104 EUR
SIAL111279-10G SIAL111279-1G SIAL111279-50G
4-Penten-1-ol, Sigma-Aldrich®
4-Penten-1-ol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 137 °C (1013 hPa)
Density: 0.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002975
CAS Number: 821-09-0
EINECS: 212-473-2
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR