cis-3-Hexen-1-ol, Sigma-Aldrich®
Supplier: Merck
Synonyms: cis-Hex-3-en-1-ol
M-Clarity™ quality level = MQ300
W256323-100G-K W256323-1KG-K W256323-4KG-K
SIALW256323-100G-KEA 154 EUR
SIALW256323-100G-K SIALW256323-1KG-K SIALW256323-4KG-K
cis-3-Hexen-1-ol, Sigma-Aldrich®
cis-3-Hexen-1-ol
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 156 °C (1013 hPa)
Melting Pt: –60 °C
Density: 0.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00063217
CAS Number: 928-96-1
EINECS: 213-192-8
UN: 1987
ADR: 3,III
Merck Index: 13,04719