trans-2-Hexen-1-ol, Sigma-Aldrich®
Supplier: Merck
Synonyms: trans-Hex-2-en-1-ol
M-Clarity™ quality level = MQ100
132667-100G 132667-25G
SIAL132667-100GEA 96.2 EUR
SIAL132667-100G SIAL132667-25G
trans-2-Hexen-1-ol, Sigma-Aldrich®
trans-2-Hexen-1-ol
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 158
Melting Pt: 65…67 °C
Density: 0.8459 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002927
CAS Number: 928-95-0
EINECS: 213-191-2
UN: 1987
ADR: 3 III