1-Butene, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

295051-100G 295051-400G
SIAL295051-100GEA 135 EUR
SIAL295051-100G SIAL295051-400G
1-Butene, Sigma-Aldrich®
1-Butene
Formula: C₄H₈
Boiling Pt: –6.3 °C (1013 hPa)
Melting Pt: –185 °C
Density: 0.6255 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009383
CAS Number: 106-98-9
EINECS: 203-449-2
UN: 1012
ADR: 2,
Merck Index: 13,01512

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR