1-Decene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
D1807-100ML D1807-1L
SIALD1807-100MLEA 82.5 EUR
SIALD1807-100ML SIALD1807-1L
1-Decene, Sigma-Aldrich®
1-Decene
Formula: C₁₀H₂₀
MW: 140.27 g/mol
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: –66 °C
Density: 0.7408 g/cm³ (20 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00009577
CAS Number: 872-05-9
EINECS: 212-819-2
UN: 3295
ADR: 3,III