1-Heptene, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
262269-100ML 262269-25ML
SIAL262269-100MLEA 259 EUR
SIAL262269-100ML SIAL262269-25ML
1-Heptene, Sigma-Aldrich®
1-Heptene
Formula: C₇H₁₄
MW: 98.19 g/mol
Boiling Pt: 93
Melting Pt: –119 °C
Density: 0.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009531
CAS Number: 592-76-7
EINECS: 209-767-8
UN: 2278
ADR: 3 II