1-Hexene, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
230545-100ML 230545-1L
SIAL230545-100MLEA 47.4 EUR
SIAL230545-100ML SIAL230545-1L
1-Hexene, Sigma-Aldrich®
1-Hexene
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 63 °C (1013 hPa)
Melting Pt: –140 °C
Density: 0.675 g/cm³ (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD00009505
CAS Number: 592-41-6
EINECS: 209-753-1
UN: 2370
ADR: 3,II