1-Hexene, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
240761-100ML 240761-50ML
SIAL240761-100MLEA 38.8 EUR
SIAL240761-100ML SIAL240761-50ML
1-Hexene, Sigma-Aldrich®
1-Hexene
Formula: C₆H₁₂
Boiling Pt: 63 °C (1013 hPa)
Melting Pt: –140 °C
Density: 0.675 g/cm³ (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD00009505
CAS Number: 592-41-6
EINECS: 209-753-1
UN: 2370
ADR: 3,II