cis-2-Hexene, Sigma-Aldrich®
Supplier: Merck
Synonyms: (Z)-2-Hexene
M-Clarity™ quality level = MQ100
538493-1G 538493-5G
SIAL538493-1GEA 48.1 EUR
SIAL538493-1G SIAL538493-5G
cis-2-Hexene, Sigma-Aldrich®
cis-2-Hexene
Formula: C₆H₁₂
Boiling Pt: 67.8 °C (1013 hPa)
Melting Pt: –141.1 °C
Density: 0.6863 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066520
CAS Number: 7688-21-3
EINECS: 231-697-1
UN: 3295
ADR: 3,II