trans-2-Hexene, Sigma-Aldrich®
Supplier: Merck
Synonyms: (E)-2-Hexene
M-Clarity™ quality level = MQ100
110892-10G 110892-1G
SIAL110892-10GEA 151 EUR
SIAL110892-10G SIAL110892-1G
trans-2-Hexene, Sigma-Aldrich®
trans-2-Hexene
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 68 °C (1013 hPa)
Melting Pt: –133 °C
Density: 0.6785 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009473
CAS Number: 4050-45-7
EINECS: 223-752-3
UN: 3295
ADR: 3,II