trans-2-Octene, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(E)-2-Octene
M-Clarity™ quality level = MQ100
111236-10G
111236-5G
SIAL111236-10GEA
162
EUR
SIAL111236-10G
SIAL111236-5G
trans-2-Octene, Sigma-Aldrich®
trans-2-Octene
Formula:
C₈H₁₆ MW: 112.22 g/mol Boiling Pt: 123 Melting Pt: –102 °C Density: 0.7199 g/cm³ (20 °C) Flash Pt: 14 Storage Temperature: Ambient |
MDL Number:
MFCD00009532 CAS Number: 13389-42-9 EINECS: 236-463-2 UN: 1993 ADR: 3 II |