trans-2-Pentene, Sigma-Aldrich®
Supplier: Merck
Synonyms: (E)-2-Pentene
M-Clarity™ quality level = MQ100
111260-10G 111260-1G 111260-5G
SIAL111260-10GEA 319 EUR
SIAL111260-10G SIAL111260-1G SIAL111260-5G
trans-2-Pentene, Sigma-Aldrich®
trans-2-Pentene
Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 37 °C (1013 hPa)
Melting Pt: –140.2 °C
Density: 0.6431 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009384
CAS Number: 646-04-8
EINECS: 211-461-4
UN: 3295
ADR: 3,II
Merck Index: 12,07263