trans-2-Methyl-2-butenal, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Trans-2-methyl-2-butenal, Tiglic aldehyde, (E)-2-Methyl-2-butenal

M-Clarity™ quality level = MQ300

W340707-100G-K W340707-1KG-K W340707-5KG-K
SIALW340707-100G-KEA 196 EUR
SIALW340707-100G-K SIALW340707-1KG-K SIALW340707-5KG-K
trans-2-Methyl-2-butenal, Sigma-Aldrich®
trans-2-Methyl-2-butenal
Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 115…117 °C (1013 hPa)
Density: 0.8688 g/cm³ (20 °C)
MDL Number: MFCD00006977
CAS Number: 497-03-0
EINECS: 207-833-0
UN: 1989
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR