3-Chloro-2-(chloromethyl)propene, Sigma-Aldrich®
Supplier: Merck
Synonyms:
3-Chloro-2-chloromethyl-1-propene, 3-Chloro-2-(chloromethyl)-1-propene
MQ100
C31104-10G
C31104-50G
SIALC31104-10GEA
161
EUR
SIALC31104-10G
SIALC31104-50G
3-Chloro-2-(chloromethyl)propene, Sigma-Aldrich®
3-Chloro-2-(chloromethyl)propene
Formula:
C₄H₆Cl₂ MW: 125 g/mol Boiling Pt: 138 Melting Pt: –11 °C Density: 1.175 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000952 CAS Number: 1871-57-4 EINECS: 217-489-3 UN: 1992 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...