3-Methyl-3-buten-1-ol, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Methylbut-3-en-1-ol, 2-Methyl-1-buten-4-ol

MQ200

129402-100ML 129402-500ML
SIAL129402-100MLEA 64.2 EUR
SIAL129402-100ML SIAL129402-500ML
3-Methyl-3-buten-1-ol, Sigma-Aldrich®
3-Methyl-3-buten-1-ol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 132
Melting Pt: –82 °C
Density: 0.853 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002933
CAS Number: 763-32-6
EINECS: 212-110-8
UN: 1987
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR