3-Methyl-2-buten-1-ol ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-Methyl-2-buten-4-ol
A16089.14
A16089.22
A16089.36
A16089.14EA
35.5
EUR
A16089.14
A16089.22
A16089.36
3-Methyl-2-buten-1-ol ≥98%
3-Methyl-2-buten-1-ol
Purity: 98+%
CAS No.: 556-82-1
Molecular Formula: C5H10O
Formula Weight: 86.13
Flash Point: 43°(109°F)
Refractive Index: 1.4440
Boiling Point: 139-140°
Beilstein Reference: 1633479
UN No.: 1987
MDL No.: MFCD00002916
Formula:
C₅H₁₀O Boiling Pt: 139…140 °C Melting Pt: –86 °C Density: 0.860 Flash Pt: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002916 CAS Number: 556-82-1 EINECS: 209-141-4 UN: 1987 ADR: 3,III |