3-Methyl-2-buten-1-ol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2-Methyl-2-buten-4-ol
M-Clarity™ quality level = MQ100
162353-100ML
162353-500ML
162353-5ML
SIAL162353-100MLEA
63.2
EUR
SIAL162353-100ML
SIAL162353-500ML
SIAL162353-5ML
3-Methyl-2-buten-1-ol, Sigma-Aldrich®
3-Methyl-2-buten-1-ol
Formula:
C₅H₁₀O MW: 86.13 g/mol Boiling Pt: 140 °C (1013 hPa) Melting Pt: –86 °C Density: 0.848 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002916 CAS Number: 556-82-1 EINECS: 209-141-4 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...