2,4,4-Trimethyl-1-pentene, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ100
443018-100ML
443018-25ML
SIAL443018-100MLEA
130
EUR
SIAL443018-100ML
SIAL443018-25ML
2,4,4-Trimethyl-1-pentene, Sigma-Aldrich®
2,4,4-Trimethyl-1-pentene
Formula:
C₈H₁₆ MW: 112.22 g/mol Boiling Pt: 102 °C (1013 hPa) Melting Pt: –93.48 °C Density: 0.71509 g/cm³ (20 °C) Flash Pt: –5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008855 CAS Number: 107-39-1 EINECS: 203-486-4 UN: 2050 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...